▸
Evaporators
▸
Evaporator Accessories
▸
Aluminum Rotor Block Holds (24) Tubes 16 x 100 mm (Set Of 2)
Aluminum Rotor Block Holds (24) Tubes 16 x 100 mm (Set Of 2)
Aluminum Rotor Block Holds (24) Tubes 16 x 100 mm (Set Of 2)
Have Questions?
Catalog number
RBA24-16-100
also known as AVL
HSN
84798200
Price (SGD)
1,202.00
Special offer
Ends:
30-Jun-2026
1,502.00
Save 300.00 (20%)
Set
Add to cart
Price (SGD)
1,202.00
Special offer
Ends:
30-Jun-2026
1,502.00
Save 300.00 (20%)
Set
Add to cart
Aluminum Rotor Block Holds (24) Tubes 16 x 100 mm (Set Of 2)
Specifications
Unit Size
Set