Filter tip Low retention 1000ul
Product Image
Filter tip Low retention 1000ul
QSP filter tips are manufactured of virgin Polypropylene with certificate of sterility included in each case
Have Questions?
Catalog NumberVolume (Metric)
QSPTFLR1121000-Q1000 μL
Catalog number QSPTFLR1121000-Q
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
1000 μL
QSP racked sterile filter tips are shrink wrapped and certified free of RNase/DNase, DNA, Pyrogen, Endotoxin, ATP, Bioburden, and PCR inhibitors. QSP filters utilize a small micron filter technology assisting in the prevention of aerosol contamination
> Low retention > Racked and sterile > Rnase and Dnase free > Universal fit for most brands of pipettes
Specifications
Volume (Metric)1000 μL
FilteredFiltered
SterilitySterile
ColorNatural
Rack ColorNatural
AutoclavableNot Autoclavable
Certifications/ComplianceISO13485
Clean ClaimsRnase Dnase free
Sterilization MethodGamma
FormatHinged Rack
Tip StyleLow Retention
Packaging10 racks of 96 tips
Quantity960
Product TypeFilter Tip
DisposableSingle-use
For Use With (Equipment)Universal Fit
GraduatedYes
Product LineQSP
MaterialPolypropylene
GraduationsYes
Unit SizePack