Check Order Status
Barnstead™ NANOpure™ Pump Assembly
Actual product may vary
Barnstead™ NANOpure™ Pump Assembly
Replacement diaphragm pump for Thermo Scientific Barnstead TII Type II Water System - Series 1403 and 1404
Have Questions?
Catalog Number
For Use With (Equipment)
PU1190X1
Type II Water System - Series 1403 and 1404
Catalog number
PU1190X1
Price (SGD)
-
For Use With (Equipment):
Type II Water System - Series 1403 and 1404
Replacement diaphragm pump for Thermo Scientific Barnstead TII Type II Water System - Series 1403 and 1404
Specifications
For Use With (Equipment)
Type II Water System - Series 1403 and 1404
Type
Water Purification Pump Assembly