Solar Power Racer Eductaion Kit
Solar Power Racer Eductaion Kit
Actual product may vary
Solar Power Racer Eductaion Kit
Construct a motorized DIY project using power from a solar cell.
Have Questions?
Catalog number PSL-5
Price (SGD)
101.00
EA
Add to cart
Price (SGD)
101.00
EA
Add to cart
Students will learn how motors and automotive transmissions work. They will also expore how solar cells convert the energy from sunlight into electricity to power the car's motor.
Requires little to no previous knowledge of electronics to construct and operate. For 4th grade and up.
Kit includes:
  • 1x Solar Cell
  • 1x Metal body
  • Gears, shafts, motor, and necessary parts
Specifications
Product TypeKit, Solar Racer
Grade4+
For Use With (Application)teaching motors, transmission, solar energy and electricity concepts
IncludesSolar cell, metal body, gears, shafts, motor, and necessary parts.
Battery TypeSolar
Unit SizeEA