Precision™ Low Temperature BOD Refrigerated Incubator
Precision™ Low Temperature BOD Refrigerated Incubator
Actual product may vary
Precision™ Low Temperature BOD Refrigerated Incubator
Precision™ Low Temperature BOD Refrigerated Incubator
Precision™ Low Temperature BOD Refrigerated Incubator
Conduct applications that require low temperatures ranging from -10°C to +60°C with these incubators, which feature microprocessor controls and are ideal for BOD.
Have Questions?
Catalog NumberCapacity (Metric)Plug TypeVoltage
PR505750R566 LNema 6-15P115 V
Catalog number PR505750R
Price (SGD)
12,792.00
EA
Add to cart
Capacity (Metric):
566 L
Plug Type:
Nema 6-15P
Voltage:
115 V
Price (SGD)
12,792.00
EA
Add to cart
Conduct applications that require temperatures ranging from -10° to +60°C with Thermo Scientific™ Precision™ Low Temperature BOD Refrigerated Incubators. Available in 2 sizes, these incubators are ideal for biochemical oxygen demand (BOD) testing, bacterial research, seed germination, fish/insect breeding and much more. Features an efficient, dependable design with microprocessor controls and forced air circulation.
Temperature Uniformity
  • Microprocessor control with easy-to-read display shows actual temperature within 0.1°C
  • Push-button controls for temperature set point selection
  • Forced-air circulation delivers excellent temperature stability
  • CFC-free, foamed polyurethane insulation prevents heat loss
  • RTD temperature probe and protected set point mode prevent accidental temperature change
Efficient Design
  • Easy-to-clean, corrosion-resistant construction
  • Compressor relay conserves energy
173L (6.1 cu. ft.) Unit
  • Economical undercounter design
  • High/low safety thermostat backups
  • Three cooling modes simplify operation: high precision with cooling; high precision without cooling; and frost-free with variable cooling
  • Four adjustable leveling feet for stable setup
  • 2A outlet easily supports apparatus inside the unit
566L (20 cu. ft.) Unit
  • Ideal for BOD applications and temperature settings at or below ambient
  • Holds over 300 BOD bottles (300mL)
  • Temperature set point selection with high- and low-temperature protection and simple calibration
  • Available with dual lamp fluorescent lighting for plant growth studies and day/night cycles, programmable lighting conditions
  • Safety relay and alarm LED alert to over/undertemperature conditions
  • Access port for independent sensors/connection of equipment inside unit
  • Cooling switch provides high temperature accuracy and saves energy at temperatures above ambient
  • RS-232 and recorder jacks for datalogging
  • Plant growth models available: PR505755L and PR505750L — with dual-lamp fluorescent lighting and programmable temperature and light cycles
Warranty: 2-year warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (English)20.0 cu. ft.
Certifications/CompliancecCSAus
Capacity (Metric)566 L
Chamber MaterialAluminum
No. of Shelves4 supplied/6 max.
Plug TypeNema 6-15P
Temperature Range (Metric)-10°C to +50°C
TypeRefrigerated Incubator
Door StyleSolid
InsulationCFC-free, foamed polyurethane
Temperature Uniformity± 0.5/0.9°C @ 37°C/20°C
Dimensions (D x W x H) Interior20 x 26.5 x 57 in. (50.8 x 67.3 x 144.8 cm)
Dimensions (L x W x H)31 x 34 x 77 in. (78.7 x 86.4 x 195.6 cm)
Height (English)77 in.
Height (Metric)1956 mm
Electrical Requirements230 V, 50/60 Hz, 3.1 A, Nema 6-15P
Voltage115 V
Amperage3.1 A
Frequency60 Hz
Unit SizeEA