PL6500 Lab Refrigerators
Actual product may vary
PL6500 Lab Refrigerators
PL6500 Lab Refrigerators
PL6500 Lab Refrigerators
PL6500 Lab Refrigerators
Store general-purpose applications in Thermo Scientific™ PL6500 Lab Refrigerators, which feature advanced alarm options to meet the requirements of a laboratory environment.
Have Questions?
Catalog NumberCapacity (Metric)
PLR386386 L
Catalog number PLR386
HSN 84185000
Price (SGD)
3,238.00
Each
Capacity (Metric):
386 L
Price (SGD)
3,238.00
Each
Store general-purpose applications in Thermo Scientific™ PL6500 Lab Refrigerators. All models feature advanced alarm options and reliable temperature stability to meet the demanding requirements of the laboratory environment.
*Available in China and limited Asia-Pacific regions only. Consult your representative for availability.
Temperature Control
  • Temperature range of 1° to 12°C (factory pre-set at 4°C)
  • Advanced air stability: ±3°C
  • Digital temperature controller with intuitive interface
  • High and low visual temperature and audio alarms
  • Automatic defrost
  • High-density, fluorine free insulation
  • Interior fan for forced air circulation (two fans on double-door model PLR1006)
Heavy-duty Construction
  • Adjustable shelves
  • Lockable, dual-pane glass doors
  • Standard casters
Other Features
  • Keyed ON/OFF switch
  • Interior lighting
  • Standard 25mm access port
Ordering Information:
Available in China and limited Asia-Pacific regions only. Consult your representative for availability.
Note:
For use in non-regulated environments.
  • Warranty varies by country, please contact us for details
  • Specifications
    Capacity (English)13.63 cu. ft.
    Capacity (Metric)386 L
    Temperature Range1°C to 12°C
    Door StyleGlass
    Shelves5
    InsulationHigh-density, fluorine free insulation
    Control TypeDigital
    DefrostAutomatic
    Monitoring OptionsAlarm
    Depth (Metric) Interior57 cm
    Height (Metric) Exterior198 cm
    Height (Metric) Interior125 cm
    Length (Metric) Exterior67 cm
    Width (Metric) Exterior66 cm
    Width (Metric) Interior52 cm
    Plug TypeChina
    Voltage220 V
    Shipping Weight (Metric)110 kg
    Unit SizeEach