PL6500 Lab Freezers
PL6500 Lab Freezers
Actual product may vary
PL6500 Lab Freezers
PL6500 Lab Freezers
PL6500 Lab Freezers
Maximize sample protection for routine laboratory storage with Thermo Scientific™ PL6500 Lab Freezers.
Have Questions?
Catalog NumberCapacity (Metric)
PLF276276 L
Catalog number PLF276
HSN 84185000
Price (SGD)
4,947.00
Special offer
Ends: 31-Mar-2026
5,138.00
Save 191.00 (4%)
Each
Add to cart
Capacity (Metric):
276 L
Price (SGD)
4,947.00
Special offer
Ends: 31-Mar-2026
5,138.00
Save 191.00 (4%)
Each
Add to cart
Maximize sample protection for routine laboratory storage with Thermo Scientific™ PL6500 Lab Freezers. Model PLF276 features six inner storage compartments. Store frequently accessed samples in the top flip-door compartments while long-term samples remain undisturbed in the bottom drawers. Advanced alarm options meet the demands of the modern laboratory environment.
Features:
  • Advanced air stability: ±3°C
  • Digital temperature controller with intuitive interface
  • High and low visual temperature and audio alarms
  • Temperature range of -20° to -40°C (factory pre-set at -40°C)
  • Manual defrost
Construction:
  • Lockable, high-quality steel door
  • Heavy-duty construction
  • Standard casters
  • High-density, fluorine free insulation
Ordering Information:
Available in China and limited Asia-Pacific and South American regions. Contact your Customer Service or Sales Representative for availability.
Specifications
Capacity (English)9.746 cu. ft.
Capacity (Metric)276 L
Temperature Range-20°C to -40°C
No. of Doors1
Door MaterialSteel
InsulationHigh-Density, Fluorine-Free
DefrostManual
Depth (Metric) Interior50 cm
Height (Metric) Exterior174 cm
Height (Metric) Interior120 cm
Length (Metric) Exterior70 cm
Width (Metric) Exterior70 cm
Width (Metric) Interior46 cm
Monitoring OptionsAlarm
Voltage220 V
Shipping Weight (Metric)123 kg
Unit SizeEach