Search Fisher Scientific
| Catalog Number | Capacity (Metric) | Depth (Metric) | Pressure |
|---|---|---|---|
| PAR4748 | 125 mL | 8.2 cm | 1900 psi |
| PAR4744 | 45 mL | 9.5 cm | 1800 psi |
For use at high temperatures and pressures.
Applications:
Dissolves both organic and inorganic analytical samples, good for digesting alumina samples or reactants of unknown behavior, useful for complete containment of volatile chemicals, safety rupture disc in bomb head blows out to release pressure through opening in cover.