General Purpose Acid Digestion Bombs
Actual product may vary
Parr
General Purpose Acid Digestion Bombs
Acid digestion bomb for organic and inorganic analytical samples
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Depth (Metric)Pressure
PAR474445 mL9.5 cm1800 psi
PAR4748125 mL8.2 cm1900 psi
Catalog number PAR4744
Price (SGD)
2,559.00
Each
Capacity (Metric):
45 mL
Depth (Metric):
9.5 cm
Pressure:
1800 psi
Price (SGD)
2,559.00
Each
For use at high temperatures and pressures.
  • Stainless-steel body
  • PTFE liner with spring-loaded closures
  • Operates at up to 250°C (482°F) and 1900psig
  • Include safety rupture discs (4749, 4744 and 4748 only)
Applications:
Dissolves both organic and inorganic analytical samples, good for digesting alumina samples or reactants of unknown behavior, useful for complete containment of volatile chemicals, safety rupture disc in bomb head blows out to release pressure through opening in cover.
Specifications
Product TypeAcid Digestion Vessel
Capacity (English)1.52 oz.
Capacity (Metric)45 mL
Depth (English)3.74 in.
Depth (Metric)9.5 cm
For Use With (Application)Dissolves both organic and inorganic analytical samples, good for digesting alumina samples or reactants of unknown behavior, useful for complete containment of volatile chemicals, safety rupture disc in bomb head blows out to release pressure through opening in cover
Height (English)5.3 in.
Height (Metric)13.5 cm
MaterialStainless Steel
Pressure1800 psi
Quantity1 Ea.
Temperature (English) Maximum482°F
Temperature (Metric) Maximum250°C
Unit SizeEach