UltraDrape™ UGPIV Barrier and Securement
Actual product may vary
Parr
UltraDrape™ UGPIV Barrier and Securement
An innovative dressing designed for use during Ultrasound-Guided Peripheral Intravenous (UGPIV) that provides dual-action barrier and securement in one.
Have Questions?
Catalog NumberPackaging
PAR341550/Box, 4 Boxes/Cs.
Catalog number PAR3415
Price (SGD)
241.00
Each
Packaging:
50/Box, 4 Boxes/Cs.
Price (SGD)
241.00
Each
UltraDrape™ is an innovative dressing designed for use during Ultrasound-Guided Peripheral Intravenous (UGPIV) that provides dual-action barrier and securement in one. A single dressing provides barrier and securement. Film layer used during ultrasound guidance is easily discarded, eliminating the need to clean transmission gel from skin.
  • UltraDrape™ UGPIV Barrier and Securement
  • UltraDrape™ is an innovative dressing designed for use during Ultrasound-Guided Peripheral Intravenous (UGPIV) that provides dual-action barrier and securement in one.
  • Sterile
  • Not made with natural rubber latex
  • A single dressing provides barrier and securement
  • Film layer used during ultrasound guidance is easily discarded, eliminating the need to clean transmission gel from skin
  • Helps reduce procedure cost when compared with
    sterile gels and covers
  • Facilitates an aseptic, no touch procedure
  • Helps minimize securement failure
  • Minimizes post-IV procedure cleanup time
Specifications
For Use With (Application)Ultrasound-Guided Peripheral Intravenous (UGPIV)
Quantity5.8 x 3.25 in. (14.72 x 8.25 cm)
Packaging50/Box, 4 Boxes/Cs.
Unit SizeEach