MB120 Moisture Analyzer
Actual product may vary
Ohaus
MB120 Moisture Analyzer
Test more samples in less time. The OHAUS™ MB120 Moisture Analyzer delivers functionality and precision that you can rely on, and is easy to operate and clean. Advanced features such as a temperature guide help to analyze optimum drying temperature, and methods and results storage allows statistical analysis of measurements.
Have Questions?
Catalog number OHAMB120
Price (SGD)
-
  • ABS top housing, stainless steel pan support
  • In-use cover, user-guide, 13 operational languages
  • Temperature Guide analyzes a sample and determines the optimal drying temperature
  • Store up to 100 drying methods for easy recall and up to 1000 total results for statistical analysis
  • Precisely controlled halogen heating dries samples uniformly, producing repeatable results of 0.01% readability
  • Four drying profiles and seven shut-off criteria help to perform customized sample tests
  • Removable pan handler and sample chamber for easy cleaning
  • Temperature Guide: Easily analyze a sample of a specific material and determine the optimum drying temperature
  • 100 Methods Library Makes Testing Faster; program the parameters for up to 100 different types of samples, then select and run your test with the push of a button
  • Store Up to 1000 Results for All Methods for Statistical Analysis; quickly retrieve data and save to flash drive
  • Three-Level User Management: Set up and lock temperature settings and method parameters with admin access, so users cant modify
  • settings
  • Touchscreen Display: Easy to read and understand
  • Icon-Driven Menu Navigation: Easy to follow, with no training required
  • 4 Mechanical Keys for Fast One-Step Operation: Tare, Start/Stop, Print, On/Off
  • USB and RS232 Ports: Quick connections to a PC or flash drive
  • Halogen heating dries samples in seconds and performs up to 40% faster than traditional infrared methods
  • The reflective sample chamber ensures uniform heating of each sample to yield reliable results of 0.01% readability
  • Features a full operating temperature range of 40–230C in 1 degree increments
  • Customize heating of specific samples with temperature control for one-step drying; just set temperature and time and begin
  • Customize your own test endpoint criteria to automatically end the test once moisture loss is no longer detected
Specifications
ModelMB120
OutputsRS232, USB device, USB host
Power120 or 240 VAC 50/60 Hz
Repeatability0.05% (3 g sample); 0.015% (10 g sample)
Readability0.01%/0.001 g
Display TypeDigital LCD
Capacity120 g
Humidity RangeMoisture Range: 0.01% to 100% (0.01% to 1000% for
Sample Size0.5 g to 120 g
Temperature Range (Metric)40°C to 230°C
TypeMoisture Analyzer
Voltage120 V
Unit SizeEach