MB120 Moisture Analyzer
Actual product may vary
MB120 Moisture Analyzer
Measures moisture content with increased precision and efficiency.
Have Questions?
Catalog NumberConnection Type
OHA30246943USB host x 2USB deviceRS232
Catalog number OHA30246943
Price (SGD)
4,437.60
Special offer
Ends: 06-Sep-2026
5,547.00
Save 1,109.40 (20%)
Each
Connection Type:
USB host x 2USB deviceRS232
Price (SGD)
4,437.60
Special offer
Ends: 06-Sep-2026
5,547.00
Save 1,109.40 (20%)
Each
  • Easily analyze specific material samples and determine the optimum drying temperature
  • 100 methods library makes testing faster
  • Store up to 1000 results for all methods for statistical analysis
  • Set up and lock temperature settings and methods parameters with admin access
  • Features wide, bright touchscreen display and intuitive menu navigation for fast, accurate moisture analysis
  • Fast infrared halogen heating technology
  • Uniform drying for precise, repeatable results
  • Flexible and powerful
  • Four drying profiles to ensure accuracy: Fast, Standard, Ramp, Step
  • Shut-Off criteria for automatic test completion
  • Features pan handler and removable parts for convenient maintenance
Specifications
ModelMB120
OutputsRS232, USB device, USB host
Repeatability0.05% (3g sample), 0.015% (10g sample)
Width (Metric) Pan90 mm
Dimensions (L x W x H)355.00 x 211.00 x 180.000 mm
Electrical Requirements120/240 VAC 50/60 Hz
Readability0.01%/0.001 g
Display TypeFull-Color 4.3 in Touchscreen WQVGA
Connection TypeUSB host x 2USB deviceRS232
Depth (Metric)355.00 mm
DescriptionMoisture Analyzer, MB120
Frequency50/60 Hz
Height (English)7.09 in.
Height (Metric)180.000 mm
Humidity RangeMoisture: 0.01% to 100%
Length (English)13.98 in.
Length (Metric)355.00 mm
Net Weight (English)11.530 lb.
Net Weight (Metric)5.230 kg
Sample Size0.500 g to 120.000 g
Temperature Range (Metric)40°C to 230°C
Voltage120 VAC or 240 VAC
Width (English)8.31 in.
Width (Metric)211.00 mm
Unit SizeEach