BioClean™ Nitrile Sterile RABS/Isolator Gloves
BioClean™ Nitrile Sterile RABS/Isolator Gloves
Actual product may vary
BioClean™ Nitrile Sterile RABS/Isolator Gloves
Sterile RABS/isolator gloves are manufactured from nitrile with low levels of particles and excellent ESD properties. Designed for use in injectable manufacturing and pharmaceutical production areas.
Have Questions?
Catalog NumberPort Size (English)Port Size (Metric)Model
NRTGGL15NIT596 in.15.2 cmGGL15NIT59
Catalog number NRTGGL15NIT59
Price (SGD)
434.00
EA
Add to cart
Port Size (English):
6 in.
Port Size (Metric):
15.2 cm
Model:
GGL15NIT59
Price (SGD)
434.00
EA
Add to cart
  • ASTM D6978-05 compliance: Cytotoxic drug and chemical resistance
  • 100% water leak tested: Assured glove integrity
  • SAL (Sterility Assurance Level) of 10-6: Ultra-clean sterile hand protection
N/A
Specifications
ASTM F739No
Certifications/ComplianceASTM D6978
Certifications/Compliance (Europe)CE 0598, EN 388:2016, EN 420:2003 + A1:2009, EN 421:2010, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 374-1, 2, 3, EN 455 Part 2, Category III
ChlorinatedYes
Cleanroom ClassClass 10 (ISO 4)
ColorWhite
Finger Thickness (English)24 mil
Finger Thickness (Metric)0.6 mm
HandAmbidextrous
Latex FreeYes
MaterialNitrile
Palm Thickness (English)20 mil
Palm Thickness (Metric)0.50 mm
Product TypeIsolator Gloves
Size9.75
Temperature (English) Max.14°F to 248°F
Temperature (Metric) Max-10°C to 120°C
Thickness (English)20 mil
Thickness (Metric)0.50 mm
For Use With (Application)Pharmaceutical Manufacturing, Laboratory Research and Development, Raw Material and Preparation, Biotechnology Manufacturing, Sterile Processing Department, Semiconductor Manufacturing
Grip DesignSmooth
Manufacturing/QMS Audit StandardsManufacturing QMS Audit Standards ISO 9001, PPE Regulation 2016 425 Module D
ModelGGL15NIT59
Port Size (English)6 in.
Port Size (Metric)15.2 cm
SeriesBIOCLEAN™ GGL36NIT59
Temperature (English) Min14°F
Temperature (Metric) Min.-10°C
Length (English)33 in.
Length (Metric)84 cm
Diameter (English) Cuff5.1 in.
Diameter (Metric) Cuff129 mm
Unit SizeEA