BioClean™ Prelude BPSL Sterile Latex Cleanroom Gloves
BioClean™ Prelude BPSL Sterile Latex Cleanroom Gloves
Actual product may vary
BioClean™ Prelude BPSL Sterile Latex Cleanroom Gloves
Powder free, sterile latex gloves offer comfort for prolonged use. Recommended for Class 10/ISO 4 cleanroom environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
NRTBPSL-80858.5
NRTBPSL-60656.5
NRTBPSL-70757.5
Catalog number NRTBPSL-8085
Price (SGD)
368.00
CS
Add to cart
Size:
8.5
Price (SGD)
368.00
CS
Add to cart
  • Natural rubber latex
  • Ambidextrous
  • Easy double-donning
  • Non-particulating EasyTear™ packaging
  • Textured surface for enhanced grip
  • Beaded cuff for stability on the arm
Specifications
Certifications/ComplianceCE 0598, Food Contact, ISO 9001:2015, ISO 9001:2008, Category III, 1.5 AQL
Certifications/Compliance (Europe)CE 0598, EN 420:2003 + A1:2009, EN 421:2010, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 455 Part 2, Food Contact, Category III
Cleanroom ClassClass 10 (ISO 4)
AQL1.5
ColorNatural
Exterior FinishFully Textured
Finger Thickness (English)7.8 mil
Finger Thickness (Metric)0.20 mm
MaterialLatex
Palm Thickness (English)6.6 mil
Palm Thickness (Metric)0.17 mm
Size8.5
SterilitySterile
Thickness (English)7.8 mil
Thickness (Metric)0.20 mm
Manufacturing/QMS Audit StandardsISO 9001:2015, ISO 9001:2008
ModelBPSL-8085
SeriesBioClean™ Prelude BPSL
Length (English)12 in.
Length (Metric)30 cm
Unit SizeCS