Nunc™ Lab-Tek™ Chambered Coverglass
Nunc™ Lab-Tek™ Chambered Coverglass
Actual product may vary
Nunc™ Lab-Tek™ Chambered Coverglass
Nunc™ Lab-Tek™ Chambered Coverglass
Nunc™ Lab-Tek™ Chambered Coverglass
Nunc™ Lab-Tek™ Chambered Coverglass
Perform high-magnification microscopy and confocal image analysis with Thermo Scientific™ Nunc™ Lab-Tek™ Chambered Coverglass.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWellsMaterialDescription
NNU1554118Borosilicate Glass (1.0)8-well Chambered Coverglass w/ non-removable wells
FIS12-565-4721Borosilicate Glass (1.0)1-well Chambered Coverglass w/ non-removable well
FIS12-565-4712Borosilicate Glass (1.0)2-well Chambered Coverglass w/ non-removable wells
NNU1553802Borosilicate Glass (1.0)2-well Chambered Coverglass w/ non-removable wells
NNU1553834Borosilicate Glass (1.0)4-well Chambered Coverglass w/ non-removable wells
FIS12-565-4708Borosilicate Glass (1.0)8-well Chambered Coverglass w/ non-removable wells
Catalog number NNU155411
Price (SGD)
-
Wells:
8
Material:
Borosilicate Glass (1.0)
Description:
8-well Chambered Coverglass w/ non-removable wells
Grow, stain and view living cells on a single device. Thermo Scientific™ Nunc™ Lab-Tek™ Chambered Coverglass with a No. 1 borosilicate glass bottom offers the optical characteristics required for high magnification and confocal image analysis. Available in 1-, 2-, 4-, and 8-well formats. The upper chamber structure cannot be removed from the coverglass.
Features:
  • Media chamber securely mounted to No. 1 borosilicate coverglass by non-toxic medical grade silicone adhesive
  • The polystyrene chamber is not removable and requires inverted microscope for viewing
  • Fitted polystyrene cover without top handle minimizes evaporation
Ordering Information:
  • Can be purchased in case of 96 or pack of 16.
  • Shipped sterile in convenient stackable incubation trays.
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Wells8
Culture Area0.8 cm2
Lid MaterialPolystyrene
Description8-well Chambered Coverglass w/ non-removable wells
Surface TreatmentCell Culture
MaterialBorosilicate Glass (1.0)
Volume (Metric) Working0.2 to 0.4 mL
Thickness (Metric)0.13 to 0.17 mm
IncludesClear polystyrene lid with handle on the side for aseptic manipulations
No. per Pack16
Dimensions (LxW)57 x 25 mm
SterilitySterile
Closure ColorClear
Unit SizePack