Nunc™ Lab-Tek™ II CC2™ Chamber Slide System
Nunc™ Lab-Tek™ II CC2™ Chamber Slide System
Actual product may vary
Nunc™ Lab-Tek™ II CC2™ Chamber Slide System
Enable cell growth directly on a glass microscope slide with the Thermo Scientific™ Nunc™ Lab-Tek™ II CC2™ Chamber Slide System.
Have Questions?
Catalog NumberWellsMaterialDescription
NNU1549174CC2 Glass (mimics polylysine)4-well Chamber Slide w/ removable wells
Catalog number NNU154917
Price (SGD)
-
Wells:
4
Material:
CC2 Glass (mimics polylysine)
Description:
4-well Chamber Slide w/ removable wells
Enable cell growth (e.g. neurons) directly on an aminated glass slide with the Thermo Scientific™ Nunc™ Lab-Tek™ II CC2™ Chamber Slide System, which features a chemically modified CC2 growth surface that mimics the characters of polylysine coating. Available in 1-, 2-, 4-, and 8-well formats with removable media chambers.
Features:
  • Growth surface provides binding sites that are optimal for fastidious cells (e.g. neurons).
  • Surface modification remains stable without refrigeration.
  • Tabbed polystyrene lid allows gas exchange with minimal evaporation.
  • Removal tool separates media chamber from glass slide.
  • Biocompatible adhesive hydrophobic well border is printed on slide.
  • Slide features a light blue frosted writing area.
  • Shelf Life: 12 months
Includes:
  • Chamber removal tool
Ordering Information:
  • Can be purchased in case of 96 or pack of 16.
  • Shipped sterile in convenient stackable incubation trays.
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Wells4
Culture Area1.7 cm2
Lid MaterialPolystyrene
Description4-well Chamber Slide w/ removable wells
Surface TreatmentCell Culture
MaterialCC2 Glass (mimics polylysine)
Volume (Metric) Working0.5 to 1.0 mL
IncludesClear polystyrene lid with handles on the top and side for aseptic manipulations
No. per Pack16
Dimensions (LxW)74 x 25 mm
SterilitySterile
Closure ColorClear
Unit SizePack