Nunc™ Glass Bottom Dishes
Nunc™ Glass Bottom Dishes
Actual product may vary
Nunc™ Glass Bottom Dishes
Using Nunc Glass Bottom Dishes in your research applications gives you the flexibility to convert your culture device into an imaging device.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
NNU15068240.4 mm
FIS12-567-40140.4 mm
Catalog number NNU150682
Price (SGD)
-
Diameter (Metric):
40.4 mm
Perform high quality imaging in the ease of a 35mm dish. The Thermo Scientific™ Nunc Glass Bottom Dish combines the convenience of a standard 35mm dish with the imaging benefits of coverglass to provide the optimum optical characteristics required for high-magnification microscopy and confocal image analysis. The interior surface of the dish bottom, including the glass area, is cell culture-treated.
Recommended for:
  • Fluorescence microscopy
  • Phase contrast microscopy
  • Confocal microscopy
  • Live cell imaging
  • Differential interference contrast microscopy
  • Fluorescence in situ hybridization (FISH)
Note:
The cover glass in the bottom cannot be detached. Do not use with methanol, ethanol, acetone or chloroform.
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure IncludedClosure Included
Closure MaterialBorosilicate Glass
DescriptionGlass Bottom Dish, 27mm
Bottom ShapeFlat
Capacity (Metric)3 mL (Well)
MaterialPolystyrene
ShapeRound
SterilitySterile
Surface TreatmentCell Culture
TypeCell Culture Dish
Volume (Metric) Working3.0 mL
IncludesLid
No. per Case20
No. per Pack1
Height (Metric)12.9 mm
Diameter (Metric)40.4 mm
Unit SizeCase