SafeKey Padlocks - Nylon with 38 mm Plastic Shackle, Keyed Different
SafeKey Padlocks - Nylon with 38 mm Plastic Shackle, Keyed Different
Actual product may vary
SafeKey Padlocks - Nylon with 38 mm Plastic Shackle, Keyed Different
SafeKey Padlocks - Nylon with 38 mm Plastic Shackle, Keyed Different
SafeKey Padlocks - Nylon with 38 mm Plastic Shackle, Keyed Different
SafeKey Padlocks - Nylon with 38 mm Plastic Shackle, Keyed Different
SafeKey Padlocks - Nylon with 38 mm Plastic Shackle, Keyed Different
SafeKey Padlocks - Nylon with 38 mm Plastic Shackle, Keyed Different
The ultimate locks for all of your safety and security needs
Have Questions?
Catalog NumberColorQuantity
NNU150265Yellow6 Pk.
Catalog number NNU150265
Price (SGD)
-
Color:
Yellow
Quantity:
6 Pk.
  • Compliments the Brady™ SafeKey Padlock range and have the same features
  • The new designed and innovative keying mechanism provides you much more unique key possibilities than a conventional standard safety padlock, without sacrificing a secure lockout
  • The unique lock mechanism is insulated and non-conductive to protect workers from shocks when key is inserted
  • The lock mechanism has 6 precision steel tumbler blades for a high number of key differences and tamper resistance
  • Locks are lightweight and resist impact, temperature, chemicals and corrosion
  • Their non-conductive, non-sparking body features a ribbed design for easier gripping and handling
  • The colour-matched key and lock body allows a fast identification in the application
  • 1 key supplied with each lock
  • Key retaining (Key won’t release until the padlock shackle is securely closed)
  • Custom keying, engraving, charting, Master Key and Grand Master Key are available
Specifications
Alternate Part NumberNYL-YLW-38PL-KD6PK
Product TypeKeyed Padlocks
Key TypeKeyed Different
ColorYellow
MaterialNylon
Diameter (English) Shackle0.25 in.
Diameter (Metric) Shackle6.4 mm
Height (English) Shackle1.5 in.
Height (Metric) Shackle38 mm
Material (Shackle)Plastic
No. of Pins6
Quantity6 Pk.
Width (English)1.5 in.
Width (English) Shackle1.3 in.
Width (Metric)38 mm
Width (Metric) Shackle33 mm
Unit SizeCase