Search Thermo Fisher Scientific
Catalog Number | Bottom Shape | Capacity (Metric) | Thread Style | Style |
---|---|---|---|---|
NNC377224 | Conical (Self-standing) | 1 mL | Internal | Writing Surface, A-R Base |
374080 | Conical (Self-standing) | 0.5 mL | Internal | No Writing Surface, A-R Base |
375353 | Conical (Self-standing) | 1 mL | External | Writing Surface, A-R Base |
FIS12-565-164N | Conical (Self-standing) | 1 mL | External | Writing Surface, A-R Base |
FKS10544641 | Conical (Self-standing) | 1 mL | External | Writing Surface, A-R Base |
NNC375353 | Conical (Self-standing) | 1 mL | External | Writing Surface, A-R Base |
FIS12-565-297 | Conical (Self-standing) | 1 mL | External | No Writing Surface, A-R Base |
366656 | Conical (Self-standing) | 1 mL | Internal | Writing Surface |
375418 | Round (Self-standing) | 1.8 mL | External | Writing Surface, A-R Base |
NNC375418 | Round (Self-standing) | 1.8 mL | External | Writing Surface, A-R Base |
347627 | Round (Self-standing) | 1.8 mL | External | No Writing Surface, A-R Base |
FKS10080601 | Round (Self-standing) | 1.8 mL | External | No Writing Surface, A-R Base |
NNC347627 | Round (Self-standing) | 1.8 mL | External | No Writing Surface, A-R Base |
377267 | Round (Self-standing) | 1.8 mL | Internal | Writing Surface, A-R Base |
FIS12-565-167N | Round (Self-standing) | 1.8 mL | Internal | Writing Surface, A-R Base |
FKS11341675 | Round (Self-standing) | 1.8 mL | Internal | Writing Surface, A-R Base |
368632 | Round (Self-standing) | 1.8 mL | Internal | Writing Surface |
FIS12-565-171N | Round (Self-standing) | 1.8 mL | Internal | Writing Surface |
FKS10674511 | Round (Self-standing) | 1.8 mL | Internal | Writing Surface |
NNC368632 | Round (Self-standing) | 1.8 mL | Internal | Writing Surface |
363401 | Round | 1.8 mL | Internal | Writing Surface |
FKS10004220 | Round | 1.8 mL | Internal | Writing Surface |
NNC363401 | Round | 1.8 mL | Internal | Writing Surface |
351934 also known as 35193 4 | Round | 1.8 mL | Internal | Writing Surface, No Cap |
379189 | Round (Self-standing) | 3.6 mL | Internal | Writing Surface, A-R Base |
FIS12-565-162N | Round (Self-standing) | 3.6 mL | Internal | Writing Surface, A-R Base |
NNC379189 | Round (Self-standing) | 3.6 mL | Internal | Writing Surface, A-R Base |
FKS10789871 | Round | 3.6 mL | Internal | Writing Surface |
337516 | Round (Self-standing) | 4.5 mL | External | Writing Surface, A-R Base |
FIS12-565-291 | Round (Self-standing) | 4.5 mL | External | Writing Surface, A-R Base |
NNC337516 | Round (Self-standing) | 4.5 mL | External | Writing Surface, A-R Base |
FIS12-565-161N | Conical (Self-standing) | 4.5 mL | Internal | Writing Surface, A-R Base |
FKS10716922 | Conical (Self-standing) | 4.5 mL | Internal | Writing Surface, A-R Base |
363452 | Round | 4.5 mL | Internal | Writing Surface |
FIS12-565-173N | Round | 4.5 mL | Internal | Writing Surface |
FKS10358131 | Round | 4.5 mL | Internal | Writing Surface |