Cryo Vial Closure Color Coders
Actual product may vary
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Nunc
Cryo Vial Closure Color Coders
Make it easier to identify specific vials quickly and simplify your inventory by using these color coders that fit into the top of cryogenic vial or tube closures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorProduct Line
NNC375868BrownNunc
354755WhiteNunc
FIS12-565-242WhiteNunc
NNC354755WhiteNunc
354879BlueNunc
NNC354879BlueNunc
354968RedNunc
NNC354968RedNunc
355018GreenNunc
NNC355018GreenNunc
355077YellowNunc
FIS12-565-246YellowNunc
NNC355077YellowNunc
355158OrangeNunc
NNC355158OrangeNunc
375868BrownNunc
375884PinkNunc
FIS12-565-252PinkNunc
NNC375884PinkNunc
375906GrayNunc
NNC375906GrayNunc
375922PurpleNunc
FIS12-565-254PurpleNunc
NNC375922PurpleNunc
375930AssortedNunc
FIS12-565-180AssortedNunc
NNC375930AssortedNunc
Catalog number NNC375868
Price (SGD)
-
Color:
Brown
Product Line:
Nunc
Make it possible to identify specific vials quickly and simplify your inventory using color coders that fit into the top of Nalgene™ Cryogenic vial closures or Nunc™ Cryogenic Tube closures.
  • Nalgene™ color coders are made from polystyrene and fit only in Nalgene™ cryogenic vial closures. Available in 5 colors
  • Nunc™ color coders are made from polycarbonate and fit only in Nunc™ cryogenic tube closures and Cryobank 2ml and 5mL tube caps. Available in 10 colors or as an assortment.
Specifications
DescriptionNunc™ CryoColor Vial Closure Coder, PS
ColorBrown
MaterialPolycarbonate
Product LineNunc
Quantity500/Pk., 2000/Cs.
Product TypeVial Closure
Unit SizePack