Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Actual product may vary
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Store samples from general cold storage to the vapor phase of liquid nitrogen with these biobanking and cell culture cryogenic tubes, available with either internal or external threads.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBottom ShapeCapacity (Metric)Thread StyleStyle
NNC375353Conical (Self-standing)1 mLExternalWriting Surface, A-R Base
374080Conical (Self-standing)0.5 mLInternalNo Writing Surface, A-R Base
375353Conical (Self-standing)1 mLExternalWriting Surface, A-R Base
FIS12-565-164NConical (Self-standing)1 mLExternalWriting Surface, A-R Base
FKS10544641Conical (Self-standing)1 mLExternalWriting Surface, A-R Base
FIS12-565-297Conical (Self-standing)1 mLExternalNo Writing Surface, A-R Base
NNC377224Conical (Self-standing)1 mLInternalWriting Surface, A-R Base
366656Conical (Self-standing)1 mLInternalWriting Surface
375418Round (Self-standing)1.8 mLExternalWriting Surface, A-R Base
NNC375418Round (Self-standing)1.8 mLExternalWriting Surface, A-R Base
347627Round (Self-standing)1.8 mLExternalNo Writing Surface, A-R Base
FKS10080601Round (Self-standing)1.8 mLExternalNo Writing Surface, A-R Base
NNC347627Round (Self-standing)1.8 mLExternalNo Writing Surface, A-R Base
377267Round (Self-standing)1.8 mLInternalWriting Surface, A-R Base
FIS12-565-167NRound (Self-standing)1.8 mLInternalWriting Surface, A-R Base
FKS11341675Round (Self-standing)1.8 mLInternalWriting Surface, A-R Base
368632Round (Self-standing)1.8 mLInternalWriting Surface
FIS12-565-171NRound (Self-standing)1.8 mLInternalWriting Surface
FKS10674511Round (Self-standing)1.8 mLInternalWriting Surface
NNC368632Round (Self-standing)1.8 mLInternalWriting Surface
363401Round1.8 mLInternalWriting Surface
FKS10004220Round1.8 mLInternalWriting Surface
NNC363401Round1.8 mLInternalWriting Surface
351934
also known as 35193 4
Round1.8 mLInternalWriting Surface, No Cap
379189Round (Self-standing)3.6 mLInternalWriting Surface, A-R Base
FIS12-565-162NRound (Self-standing)3.6 mLInternalWriting Surface, A-R Base
NNC379189Round (Self-standing)3.6 mLInternalWriting Surface, A-R Base
FKS10789871Round3.6 mLInternalWriting Surface
337516Round (Self-standing)4.5 mLExternalWriting Surface, A-R Base
FIS12-565-291Round (Self-standing)4.5 mLExternalWriting Surface, A-R Base
NNC337516Round (Self-standing)4.5 mLExternalWriting Surface, A-R Base
FIS12-565-161NConical (Self-standing)4.5 mLInternalWriting Surface, A-R Base
FKS10716922Conical (Self-standing)4.5 mLInternalWriting Surface, A-R Base
363452Round4.5 mLInternalWriting Surface
FIS12-565-173NRound4.5 mLInternalWriting Surface
FKS10358131Round4.5 mLInternalWriting Surface
Catalog number NNC375353
Price (SGD)
-
Bottom Shape:
Conical (Self-standing)
Capacity (Metric):
1 mL
Thread Style:
External
Style:
Writing Surface, A-R Base
Store samples from general cold storage to the vapor phase of liquid nitrogen with Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes, available with either internal or external threads.
Features:
  • Non-cytotoxic, non-mutagenic raw materials and pyrogen-free IVD compatible tubes for added sample security
  • Certified SAL of 10-6 for the highest sterility assurance available
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
  • Bulk, uncapped, non-sterile versions available for less demanding applications
Specifications
Diameter (Metric)12.5 mm
Product TypeCryogenic Storage Tube
DescriptionWith writing surface
Diameter (English) Outer0.492 in.
AutoclavableNot Autoclavable
Capacity (Metric)1 mL
Bottom ShapeConical (Self-standing)
MaterialPolypropylene
Thread StyleExternal
Length (English)1.614 in.
Length (Metric)41 mm
No. per Pack500
StyleWriting Surface, A-R Base
RNase FreeYes
Vapor LN2 (to -150°C)Yes
Product LineNunc™
SterilitySterile
Anti-RotationYes
Packaging Quantity500/Pk.
Closure MaterialPolyethylene
Capacity (English)0.0338 oz.
Dnase FreeYes
Closure TypeScrew Cap
PrintedYes
Unit SizePack