Storage Tube Protection
Actual product may vary
Storage Tube Protection
Storage Tube Protection
Storage Tube Protection
Nunc
Storage Tube Protection
Give your cryogenic storage tubes and vials extra protection, such as when shipping or storing in liquid or vapor phase nitrogen with these storage accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeQuantity
NNC343958Tube Wrap15/Pk., 300/Cs.
343958Tube Wrap15/Pk., 300/Cs.
534479Cryo Storage Box350/Pk., 350/Cs.
534592
also known as PA534592
Cryo Storage Box200/Pk., 200/Cs.
FKS10461482Cryo Storage Box200/Pk., 200/Cs.
Catalog number NNC343958
Price (SGD)
-
Product Type:
Tube Wrap
Quantity:
15/Pk., 300/Cs.
Give your cryogenic storage tubes and vials extra protection when needed with Thermo Scientific™ Storage Tube Protection accessories. For example, safeguard your samples when storing in liquid or vapor phase nitrogen, or when shipping samples. These cryogenic storage accessories, such as tube wraps, sleeves and mini-boxes, have safe handling in mind.
  • If you want to submerge your cryotubes in the liquid phase of nitrogen, seal them first inside Nunc™ Cryoflex™ tubing to prevent liquid nitrogen from infiltrating the tube. Cryoflex™ tubing can used to crimp and heat shrink an impenetrable safety skin around the tube prior to submersion.
  • Nalgene™ CryoSleeves™ slide over any Nunc or Nalgene Cryocane and prevent tubes from falling out of the cryocane and into the bottom of the liquid nitrogen dewar. Nalgene™ CryoSleeves™ do not prevent infiltration of liquid nitrogen. Cryogenic tubes mounted in the cane must first be sealed inside Cryoflex™ tubing prior to applying the CryoSleeve™ if liquid phase submersion is required.
  • Nunc CryoTube™ miniboxes with a friction-fit lid are ideal for storing small quantities of samples and shipping.Made with durable, high-impact, polystyrene to protect samples.
Specifications
DescriptionNunc CryoFlex Tube Wrap, 500 mm, 14 mm ID
Diameter (Metric) Inner14 mm
Quantity15/Pk., 300/Cs.
Height (Metric)500 mm
Product TypeTube Wrap
Unit SizePack