Search Fisher Scientific
| Catalog Number | Volume | Quantity |
|---|---|---|
| NNC254437 | 10 μL | Carton of 600 |
Easily identify color-coded Thermo Scientific™ Nunc™ Disposable Loops and Needles, precision molded from nontoxic (USP Class VI) polystyrene material to provide a smooth loop surface. Thermo Scientific Nunc products are high quality plastics for use in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits.
Features: