Nunc™ Serological Pipettes
Nunc™ Serological Pipettes
Actual product may vary
Nunc™ Serological Pipettes
Nunc™ Serological Pipettes
Nunc™ Serological Pipettes
Choose size and packaging options to advance your research with complete confidence. Did you know we now offer NEW Serological Pipettes that offer higher quality and cost savings?
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Packaging Type
NNC17035610 mLIndividual Paper Wrap
FIS12-567-6001 mLIndividual Paper/Plastic Wrap
NNC1703542 mLIndividual Paper/Plastic Wrap
FIS12-567-60310 mLIndividual Paper Wrap
FKS1342477625 mLIndividual Paper/Plastic Wrap
FIS12-567-60910 mLIndividual Plastic Wrap
FIS12-567-61025 mLIndividual Plastic Wrap
FKS1349029725 mLBulk packed, 25/bag
FIS12-567-61910 mLIndividual Paper/Plastic Wrap
Catalog number NNC170356
Price (SGD)
-
Volume (Metric):
10 mL
Packaging Type:
Individual Paper Wrap
Select size and packaging options to advance your research with complete confidence when using Thermo Scientific™ Nunc™ Serological Pipettes. These accurate, disposable plastic pipettes are plugged and sterilized.
Features:
  • 100% virgin polystyrene for maximum clarity
  • Certified non-pyrogenic
  • Non-cytotoxic
  • Sterilized pipettes are Gamma irradiated at a validated dose to ensure Sterility Assurance Level of 10-6
  • Bold clear graduations with easy-to-read scale markings
  • Non-dripping tip for pipette precision
  • Color-coded packaging for ease in sorting and selecting the correct size
  • Individually wrapped; paper/plastic or plastic film; peel or pop through for easy opening
  • Bulk packed for frequent use; reduces packaging waste
  • Shortie pipette with ergonomically friendly design for use under laminar hood
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Aspirator PlugYes
MaterialPolystyrene
SterilitySterile
Quantity50/Pk., 200/Cs.
Packaging TypeIndividual Paper Wrap
Volume (Metric)10 mL
Unit SizeCase