Nunc™ Non-treated Flasks
Nunc™ Non-treated Flasks
Actual product may vary
Nunc™ Non-treated Flasks
Minimize the chance of contamination with these high-quality, optically clear polystyrene Thermo Scientific™ Nunc™ Non-treated Flasks for suspension cell cultures.
Have Questions?
Catalog NumberCulture Area
NNC159926175 cm²
Catalog number NNC159926
Price (SGD)
-
Culture Area:
175 cm²
Benefit from the user-friendly designs of these high-quality, optically clear polystyrene Thermo Scientific™ Nunc™ Non-treated Flasks for suspension cell cultures.
Features:
  • White caps for easy identification
  • The ergonomic design of Thermo Scientific™ EasyFlask™ allows for easy handling and pipetting
  • Thermo Scientific™ TripleFlask™ with three parallel layers has the same external dimensions of a standard 175 cm2 flask
  • The filter cap ensures constant airflow while minimizing chances of contamination
  • Sterilie and non-pyrogenic
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure ColorWhite
DescriptionNunc Non-treated T175 EasYFlask, Filter Cap
MaterialPolystyrene
Neck StyleAngled
No. per Case30
No. per Pack5
SterilitySterile
Closure TypeFilter
Culture Area175 cm²
Surface TreatmentNon-treated
Volume (Metric) Working55 mL
Unit SizeCase