SafeKey Padlocks - aluminum with 76 mm Steel Shackle, Keyed Alike
SafeKey Padlocks - aluminum with 76 mm Steel Shackle, Keyed Alike
Actual product may vary
SafeKey Padlocks - aluminum with 76 mm Steel Shackle, Keyed Alike
SafeKey Padlocks - aluminum with 76 mm Steel Shackle, Keyed Alike
SafeKey Padlocks - aluminum with 76 mm Steel Shackle, Keyed Alike
SafeKey Padlocks - aluminum with 76 mm Steel Shackle, Keyed Alike
The ultimate locks for indoor and outdoor heavy duty safety applications.
Have Questions?
Catalog NumberColorQuantity
NNC150239Yellow3 Pk.
Catalog number NNC150239
Price (SGD)
-
Color:
Yellow
Quantity:
3 Pk.
  • Manufactured with solid aluminum body and steel shackle, Brady™'s padlocks are lightweight and rugged
  • Compliments the Brady™ SafeKey Padlock range and have the same features
  • Locks are chemical and corrosion resistant
  • The unique lock mechanism is insulated and has 6 precision steel tumbler blades for a high number of key differences and tamper resistance
  • The colour-matched key and lock body allows a fast identification in the application
  • 1 key is supplied with each lock
  • Custom keying, engraving, charting, Master Key and Grand Master Key are available
Specifications
Alternate Part NumberALU-YLW-76ST-KA3PK
Product TypeKeyed Padlocks
Key TypeKeyed Alike
ColorYellow
MaterialAnodized Aluminum
Diameter (English) Shackle0.25 in.
Diameter (Metric) Shackle6.4 mm
Height (English) Shackle3 in.
Height (Metric) Shackle76 mm
Material (Shackle)Steel
No. of Pins6
Quantity3 Pk.
Width (English)1.5 in.
Width (English) Shackle1.3 in.
Width (Metric)38 mm
Width (Metric) Shackle33 mm
Unit SizeCase