Perform suspension cell cultures with Thermo Scientific™ Nunc™ Non-Treated Multidishes.
Have Questions?
Change view
Catalog Number
No. of Wells
NNC150200
12
FKS10559532
12
FIS12-566-80
6
3 Options
Catalog number NNC150200
Price (SGD)
-
No. of Wells:
12
Perform suspension cell cultures with Thermo Scientific™ Nunc™ Non-Treated Multidishes.
Flat-bottom round wells
Excellent optical properties
Raised well rims to lower the risk of cross-contamination
Lid with notched corners for good fit and easy orientation
Sterile and non-pyrogenic
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.