Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom: Sterile
Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom: Sterile
Actual product may vary
Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom: Sterile
Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing and storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeClosure Type
NLG4116-28002800 mL70 mmVented
FIS10-529125 mL38-430 mmVented
FIS10-530250 mL38-430 mmVented
FIS10-531500 mL45-430 mmVented
FKS10082332500 mL45-430 mmVented
Catalog number NLG4116-2800
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
2800 mL
Closure Size:
70 mm
Closure Type:
Vented
Save preparation time and avoid contamination risk by using Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Features:
  • Sterile and single use to reduce contamination risk
  • BPA-free
  • Collapses when autoclaved to reduce bio hazardous waste
  • Graduated for quick volume assessment
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (English)95 oz.
Capacity (Metric)2800 mL
MaterialPETG
No. per Case4
No. per Pack1
ShapeFernbach
Surface CoatingNone
AutoclavableNot Autoclavable
DisposableSingle-use
GraduatedYes
Mouth StyleNarrow
SterilitySterile
Bottom ShapeBaffled
Closure TypeVented
Closure Size70 mm
Unit SizeCase