Nalgene™ Polypropylene Graduated Cylinders
Actual product may vary
Thermo Scientific
Nalgene™ Polypropylene Graduated Cylinders
Conduct applications accurately with these graduated cylinders, which feature generous pour spouts and molded-in graduations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
NLG3662-002525 mL0.5 mL
3662-001010 mL0.2 mL
FIS08-572A10 mL0.2 mL
3662-002525 mL0.5 mL
FIS08-572B25 mL0.5 mL
FIS08-572C50 mL1 mL
FIS08-572D100 mL1 mL
FKS10683931100 mL1 mL
FIS08-572E250 mL2 mL
FIS08-572F500 mL5 mL
3662-10001000 mL10 mL
FIS08-572G1000 mL10 mL
FKS104539211000 mL10 mL
FIS08-572H2000 mL20 mL
FIS08-572J4000 mL50 mL
Catalog number NLG3662-0025
Price (SGD)
-
Capacity (Metric):
25 mL
Graduations:
0.5 mL
Conduct applications accurately with Thermo Scientific™ Nalgene™ Polypropylene Graduated Cylinders, which feature generous pour spouts, molded-in graduations and a large blue base. There is no meniscus to confuse readings, eliminating guesswork; the cylinders can be chemically sterilized and are suitable for food and beverage use. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
Features:
  • Excellent chemical resistance for a wide variety of solutions
  • Resists temperatures up to 135°C (275°F) for a variety of uses
  • Autoclavable (autoclaving will affect accuracy)
  • Molded-in graduations are silk-screened for easy reading (10mL to 1L sizes only)
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
Ordering Information:
Individually packaged.
Recycling Information: Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Thermo Fisher Scientific encourages recycling Nalgene polypropylene labware at end of product life in communities where polypropylene can be recycled.
Nalgene beakers and graduated cylinders are delivered in corrugate cartons made from 100% post-consumer recycled material.  Packaging bags are made from LDPE and are readily recyclable once the label is removed.   Please continue the cycle and recycle your cartons and bags.
.
Specifications
Capacity (Metric)25 mL
MaterialPolypropylene
Tolerance±0.34 mL
Certifications/ComplianceCFR 21 Part 177.1520, ISO 6706, ASTM E1272
Scale1.5mL to 25mL in 0.5mL increments
IncludesLarge Blue Base
Graduations0.5 mL
AutoclavableAutoclavable
Limit of Error±0.34mL
ShapeRound
Max. Temperature (English)250°F
For Use With (Application)Food and Beverage Use
Base TypeOctagonal
Height (English)5.81 in.
Height (Metric)147.5 mm
Product TypeGraduated Cylinder
Max. Temperature (Metric)121°C
Unit SizePack