Nalgene™ Wide-Mouth EP Tox/TCLP Bottle Made With Teflon™ FEP, includes PFA-lined Closure
Nalgene™ Wide-Mouth EP Tox/TCLP Bottle Made With Teflon™ FEP, includes PFA-lined Closure
Actual product may vary
Thermo Scientific
Nalgene™ Wide-Mouth EP Tox/TCLP Bottle Made With Teflon™ FEP, includes PFA-lined Closure
Thermo Scientific™ Nalgene™ Wide-Mouth Teflon™ EP Tox/TCLP Bottles can be used at high and low temperatures for trace metal analysis and applications with organic solvents.
Have Questions?
Catalog NumberCapacity (Metric)
NLG2101-22002.2 L
Catalog number NLG2101-2200
Price (SGD)
-
Capacity (Metric):
2.2 L
Use Thermo Scientific™ Nalgene™ Wide-Mouth EP Tox/TCLP Bottle made with Teflon™ FEP with PFA-lined Closure at high and low temperatures for trace metal analysis and applications with organic solvents. Offering laboratories a safer alternative without sacrificing needed accuracy, these bottles are specially designed for US EPA Method 1311: Toxicity Characteristic Launching Program (TCLP).
  • Temperature-resistant PTFE resin FEP bottle has wide mouth for testing large-sized samples
  • Autoclavable
  • Leakproof†
  • PTFE resin-lined polypropylene caps included
Notes:
Completely disengage threads before autoclaving.
These tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes.
Specifications
Capacity (Metric)2.2 L
ColorTranslucent
DescriptionWide Mouth EP Tox/TCLP Bottle
Diameter (English) Inner3.5 in.
Diameter (English)4.7 in.
Diameter (Metric) Inner89 mm
For Use With (Application)Storage, Transportation ​
MaterialTeflon FEP
No. per Case2/Cs.
No. per Pack1 Ea.
SterilityNon-sterile
Closure ColorWhite
Certifications/ComplianceSpecifically designed for U.S. EPA Method 1311: Toxicity Characteristic Leaching Program (TCLP)
AutoclavableAutoclavable
MouthWide
ShapeRound
Height (Metric)241 mm
Height (English)9.5 in.
Closure LinerTeflon
Closure MaterialPolypropylene
Closure Size100 mm
Diameter (Metric)119 mm
Closure TypeScrew Cap
Unit SizeCase