Multidrop™ Combi SMART Reagent Dispenser Accessories
Multidrop™ Combi SMART Reagent Dispenser Accessories
Actual product may vary
Multidrop™ Combi SMART Reagent Dispenser Accessories
Guarantee excellent dispensing results and day-to-day performance with reagent dispenser accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionVolume (Metric)Quantity
N15137SMART 2 Standard, New RFID Tag5 to 2500 μL1 Ea.
MIBN15133SMART 2 Small Plastic Tip, New RFID Tag0.5 to 50 μL1 Ea.
MIBN15135SMART 2 Small Metal Tip, New RFID Tag0.5 to 50 μL1 Ea.
Catalog number N15137
Price (SGD)
-
Description:
SMART 2 Standard, New RFID Tag
Volume (Metric):
5 to 2500 μL
Quantity:
1 Ea.
Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi SMART Reagent Dispenser Accessories.
  • Designed for use with Multidrop Combi SMART+ dispensers
  • Equipped with radio frequency identification (RFID) tags to store lifetime dispensing data and display remaining usage time
  • Available in standard and small tube versions
  • Distinguished from normal cassettes by antenna tags
  • Cat. No. N15133 to N15139 are to be used with the Multidrop Combi with SMART2 option instrument with serial numbers 836-2846SM onwards
  • Cat. No. 24073292, 24073001, 24073297, 24073002, 24072675, 24072678 and 24072676 are to be used with the Multidrop Combi with SMART option instrument with serial numbers up to 836-2845SM
Specifications
Product TypeTube Dispensing Cassette
AutoclavableAutoclavable (50 times at 121°C, 1 bar, 20 min.)
DescriptionSMART 2 Standard, New RFID Tag
MaterialPolypropylene Tip, Silicone Tubing
Volume (Metric)5 to 2500 μL
For Use With (Equipment)Multidrop Combi with SMART2 option instrument with serial numbers 836-2846SM onwards
Quantity1 Ea.
Volume (Metric) Dead<7 mL
Diameter (Metric)Tubing: 1.3 mm; Tip: 0.5 mm
Unit SizeEA
Frequently asked questions (FAQs)
Can the SMART cassettes with RFID tags be used with the standard Multidrop Combi without SMART option or vice versa?
Yes, the cassettes are interchangeable, however, the calibration data cannot be viewed and the information about the cassette is not updated. Also standard cassettes can be used with the Multidrop Combi with SMART option.
What should I do if the liquid foams during dispensing with the Multidrop Combi?
Some liquids foam more easily than others. One option is to reduce the pump rotor speed. There are three dispensing speeds available: High, Med (medium), and Low.
What materials come into contact with liquids to be dispensed by the Multidrop Combi?
The Small tube plastic tip dispensing cassette, Cat. No. 24073290, and the Standard tube dispensing cassette, Cat. No. 24072670, are made of: PEEK, silicone, and PP (polypropylene).
The Small tube metal tip dispensing cassette, Cat. No. 24073295, is made of: PEEK, silicone, ruby, and stainless steel.
Do particles in the reagent affect dispensing with the Multidrop Combi?
Possibly. It is recommended that you ensure that there are no particles greater than 50 μm in the reagent to avoid blockage of the tips, especially when working with small tube cassettes.
What is the tip inner diameter (ID) of the dispensing cassettes of the Multidrop Combi?
The tip inner diameter (ID) is either 0.22 mm (Small tube plastic tip dispensing cassette, Cat. No. 24073290, and Small tube metal tip dispensing cassette, Cat. No. 24073295), or 0.5 mm (Standard tube dispensing cassette, Cat. No. 24072670).