TWEEN 20, MP Biomedicals™
TWEEN 20, MP Biomedicals™
TWEEN 20, MP Biomedicals™
TWEEN 20, MP Biomedicals™
MP Biomedicals
TWEEN 20, MP Biomedicals™
TWEEN 20, Molecular biology reagent, Viscous pale yellow liquid
Have Questions?
Catalog NumberQuantity
MPB11TWEEN201500 mL
Catalog number MPB11TWEEN201
Price (SGD)
129.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
129.00
EA
Add to cart
Specifications
ColorPale Yellow
CAS9005-64-5
Physical FormLiquid
Boiling Point>100°C (literature)
Storage Note 1Store at Room temperature(15-30°C).
View more
  • TWEEN 20 is a nonionic detergent widely used in biochemical applications, such as emulsifying agents for the preparation of stable oil-inwater emulsions.
  • It has been used in preextraction of membranes to remove peripheral proteins.
  • TWEEN 20 has been used as a blocking agent for membrane based immunoassays at a typical concentration of 0.05%.
  • TWEEN 20 can be used for lysing mammalian cells at a concentration of 0.05 to 0.5%. TWEEN 20 is a polyoxyethylene sorbitol ester.