TRIS, MP Biomedicals™
TRIS, MP Biomedicals™
TRIS, MP Biomedicals™
MP Biomedicals
TRIS, MP Biomedicals™
TRIS, Molecular biology grade, >99.8%, White Crystalline Powder
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Catalog number MPB11TRIS0500
Price (SGD)
169.00
Each
Quantity:
500 g
Request bulk or custom format
Price (SGD)
169.00
Each
Specifications
CAS77-86-1
Melting Point167°C to 172°C (literature)
Boiling Point219°C to 220°C/10 mmHg(literature)
ColorWhite
Storage Note 1Store at Room temperature(15-30°C), desiccate.
View more
  • Tris may be used by itself as a buffer or as a component of mixed buffer formulations e.g. reaction buffers for restriction enzymes or nucleic acid modifying enzymes.
  • Tris salts are used in protein crystallization at various pH values.
  • The use of low-ionic strength Tris buffers in the formation of intermediate filaments of lamin from Caenorhabditis elegans has been described.
  • Tris has been utilized in studies of double stranded complexes of peptide nucleic acids (PNA) and their complementary DNA sequences.
  • The use of Tris in capillary electrochromatography and UV analysis of tocopherols and tocotrienols has been reported.
  • Tris is an established basimetric standard and buffer used in biochemistry and molecular biology