TAE 50X BUFFER, MP Biomedicals™
MP Biomedicals
TAE 50X BUFFER, MP Biomedicals™
TAE 50X BUFFER
Have Questions?
Catalog NumberQuantity
MPB11TAE50X011 L
Catalog number MPB11TAE50X01
Price (SGD)
326.00
Each
Quantity:
1 L
Request bulk or custom format
Price (SGD)
326.00
Each
Specifications
ColorClear Colorless
Storage Note 1Store at 2-8°C
Physical FormSolution
Quantity1 L
FormulationTris Base: 2M; Acetic Acid : 1M; EDTA : 0.05 M Acetic acid: 1M EDTA: 0.05M
View more
  • TAE running buffer is the most commonly used buffer for DNA agarose gel electrophoresis but is also used for non-denaturing RNA agarose gel electrophoresis.
  • Buffer for Electrophoresis.
  • Prepare electrophoresis buffers