Recommended for sharp resolution of nucleic acid fragments from 400 to 8000bp. Agarose Standard, MP Biomedicals is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae (electroendoosmosis value 0.12), making it an essent
Have Questions?
Catalog Number
Quantity
MPB11AGAH0500
500 g
Catalog number MPB11AGAH0500
Price (SGD)
2,248.00
Each
Quantity:
500 g
Request bulk or custom format
Price (SGD)
2,248.00
Each
Specifications
CAS9012-36-6
Melting Point∼89°C (1.5% gel)
Recommended StorageRoom temperature (15-30°C).
Quantity500 g
Gel Strength≥1200g/cm3
View more
This Standard Agarose may be used at concentrations between 0.8% and 2%. Southern and Northern transfers can be performed efficiently from them.