RNAPro Solution
RNAPro Solution
Actual product may vary
MP Biomedicals
RNAPro Solution
RNAPro Solution is a proprietary lysis buffer included with all of the FastRNA Pro purification kits. It can be used with the kits or with any other purification protocol as the primary lysis buffer to isolate total RNA from any type of plant or animal tissue, cultured cells, microbes, or soil and environmental samples.
Have Questions?
Catalog NumberQuantity
MPB1160550502 x 27.5 mL
Catalog number MPB116055050
Price (SGD)
159.00
EA
Add to cart
Quantity:
2 x 27.5 mL
Price (SGD)
159.00
EA
Add to cart
  • Extracted RNA is suitable for RT-PCR analysis and other downstream applications
  • Using the FastPrep-24™ or FastPrep™ FP120 Instrument, samples can be homogenized by using MP Lysing Matrix tubes
  • Use with FastPrep-24 or FastPrep-96 (with ConeFlex Legacy Adapter), or with optional CoolPrep Adapter kits to lyse, grind or homogenize samples, for the extraction and purification of RNA
Research Use Only (RUO). Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Specifications
Sample TypeSeeds, Spores, Bacteria (Gram +/-), Animal Tissues, Feces, Food, Fungi, Plants, Roots, Soil, Wastewater, Yeast
For Use With (Equipment)FastPrep-24, FastPrep 120, FastPrep-96, SuperFastPrep-1
For Use With (Application)Isolation of RNA from most biological or environmental samples
Product TypeRNAPro Solution
Quantity2 x 27.5 mL
Sample VolumeScalable
Unit SizeEA
Contents & Storage
Room Temperature