Empty FastPrep™ 2 mL Lysing Matrix Tubes
Empty FastPrep™ 2 mL Lysing Matrix Tubes
Actual product may vary
MP Biomedicals
Empty FastPrep™ 2 mL Lysing Matrix Tubes
Available empty for those who wish to fill their own tubes. Empty FastPrep™ 2 mL Lysing Matrix Tubes (non-skirted, no caps) are 2 mL tubes that work with FastPrep or other bead-beater instruments to lsye, grind or homogenize samples.
Have Questions?
Catalog NumberVolume (Metric)
MPB1150762002 mL
Catalog number MPB115076200
Price (SGD)
284.00
EA
Add to cart
Volume (Metric):
2 mL
Price (SGD)
284.00
EA
Add to cart
  • Used in FastPrep systems.
  • Add sample to empty tube, add appropriate Lysing Matrix material and lysing buffer, and apply cap.
  • Empty FastPrep 2 mL tubes are screw cap tubes and can be used with any color cap.
  • They are designed for grinding and lysing samples with FastPrep instruments.
  • Non-skirted
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Specifications
ColorClear
Volume (Metric)2 mL
Storage RequirementsRoom Temperature (15°C to 30°C)
Quantity500 x 2 mL
Product TypeEmpty Lysing Matrix Tubes
Unit SizeEA