CSM-HIS, Powdered Media
CSM-HIS, Powdered Media
Actual product may vary
MP Biomedicals
CSM-HIS, Powdered Media
Single dropout of Complete Supplement Mixture (CSM) of amino acids for S. cereviseae growth media
Have Questions?
Catalog NumberQuantity
MPB11451031210 g
Catalog number MPB114510312
Price (SGD)
225.00
EA
Add to cart
Quantity:
10 g
Price (SGD)
225.00
EA
Add to cart
Formulation
  • Add 0.77g of CSM-His to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes
Applications:
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Certifications/ComplianceMolecular biology Certified™
FormPowder
Product TypeYeast Media
FormulationAdenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 0mg/L, L-Isoleucine 50mg/L, L-Leucine 100mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 20mg/L, L-Valine 140mg/L
For Use With (Application)For growth of Saccharomyces cerevisiae
Quantity10 g
Shelf Life4 Years
Unit SizeEA
Contents & Storage
15°C to 30°C