YNB, Powdered Media: Without Ammonium Sulfate
YNB, Powdered Media: Without Ammonium Sulfate
Actual product may vary
MP Biomedicals
YNB, Powdered Media: Without Ammonium Sulfate
Can be prepared by adding a carbon source, the appropriate amino acids, and agar if necessary
Have Questions?
Catalog NumberQuantity
MPB114027012227 g
Catalog number MPB114027012
Price (SGD)
249.00
EA
Add to cart
Quantity:
227 g
Price (SGD)
249.00
EA
Add to cart
  • Nitrogen-based yeast
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Certifications/ComplianceMolecular Biology Certified™
FormPowder
Product TypeMicrobiology Media
FormulationNitrogen Source: Ammonium Sulfate 5000mg/L; Vitamins: Biotin 0.002mg/L, Calcium Pantothenate 0.4mg/L, Folic Acid 0.002mg/L, Inositol 2mg/L, Niacin 0.4mg/L, P-Aminobenzoic Acid 0.2mg/L, Pyridoxine Hydrochloride 0.4mg/L, Riboflavin 0.2mg/L, Thiamine Hydrochloride 0.4mg; Trace Elements: Boric Acid 0.5mg, Copper Sulfate 0.04g, Potassium Iodide 0.1mg, Ferric Chloride 0.2mg, Manganese Sulfate 0.4mg, Sodium Molybdate 0.2mg, Zinc Sulfate 0.4mg; Salts: Magnesium Sulfate 500mg, Sodium Chloride 100mg, Potassium Phosphate monobasic 1000mg, Calcium Chloride 100mg
For Use With (Application)For vigorous growth of Saccharomyces cerevisiae
Quantity227 g
Unit SizeEA
Contents & Storage
15°C to 30°C