M9 Minimal Medium Salts (Maniatis Formulation), Powdered
M9 Minimal Medium Salts (Maniatis Formulation), Powdered
Actual product may vary
MP Biomedicals
M9 Minimal Medium Salts (Maniatis Formulation), Powdered
Used for preparing a defined synthetic medium for the growth of E. coli host
Have Questions?
Catalog NumberQuantity
MPB1130370321000 g
Catalog number MPB113037032
Price (SGD)
558.00
EA
Add to cart
Quantity:
1000 g
Price (SGD)
558.00
EA
Add to cart
  • A commonly used base for preparing a defined synthetic medium for the growth of most E. coli host cells
  • Powder is reconstituted in sterile purified water, (or water with melted purified agar added)
  • Additional nutrients, such as glucose or other carbon source, casamino acids, vitamins, or antibiotics are added when the solution cools
  • Add additional nutrients before plates are poured or media is inoculated with bacteria
Specifications
Certifications/ComplianceMolecular Biology Certified
Concentration56.4 g/L of a 5X Solution
DescriptionA 56.4 g sample of M9 Minimal Salts was dissolved in an aqueous solution to 1 liter, the pH adjusted to 7.4 (if needed), and the solution autoclaved to make a sterile 5X concentrate
FormPowder
Media TypeM9 Minimal Medium
Product TypeMicrobiology Media
SterilitySterile
Formulation33.9 g Na2HPO4; 15 g KH2PO4; 2.5 g NaCl; 5 g NH4Cl
For Use With (Application)Preparing a Defined Synthetic Medium for The Growth of E. Coli Host
pH6.8
Quantity1000 g
Shelf LifeExpiration Date
Shipping ConditionAmbient
SolubilitySoluble Aqueous Solution
Specimen TypeBacterial
Strain DesignationsE. coli
Unit SizeEA
Contents & Storage
15° to 30°C