SOC (Powder)
SOC (Powder)
Actual product may vary
MP Biomedicals
SOC (Powder)
Maximizes the transformation efficiency of competent cells. MP Biomedicals SOC (Powder) is a special medium for incubating competent cells in the recovery step of E. coli cell transformations. It allows expression of transferred resistance genes before ex
Have Questions?
Catalog number MPB113031012
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Chosen reagents are combined in the accurate proportions according to the formula and thoroughly blended for a uniform distribution. A sample of the blended formulation is used to prepare liquid. The combinations are mixed to dissolve components and then autoclaved at 121°C for 15 minutes.
Specifications
Certifications/ComplianceMolecular Biology Certified™
DescriptionSOC
FormPowder
FormulationPer liter: 20 g Tryptone-B; 5 g Yeast Extract-B; 0.5 g NaCl; 5 g MgSO4-7H2O; 20 mM glucose. Small quantities of biological buffers have been added for proper pH adjustment.
For Use With (Application)Recovery step of Escherichia coli competent cell transformations
Quantity227 g
Regulatory StatusRUO
Specimen TypeE. coli
Unit SizeEA
Contents & Storage
Store at room temperature.