LB-Agar, Large Capsules
LB-Agar, Large Capsules
Actual product may vary
MP Biomedicals
LB-Agar, Large Capsules
Contents/liter: LB Medium, 15g Agar-B
Have Questions?
Catalog NumberQuantity
MPB1130022465000 g
Catalog number MPB113002246
Price (SGD)
3,314.00
EA
Add to cart
Quantity:
5000 g
Price (SGD)
3,314.00
EA
Add to cart
Use 8 large capsules/L.
Specifications
Certifications/ComplianceMolecular Biology Certified™
FormatCapsule
GradeMolecular Biology
Product TypeAgar
FormulationTryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L, Agar 15g/L
For Use With (Application)For growth of bacteria
Quantity5000 g
Shelf Life4 Years
Unit SizeEA
Contents & Storage
15°C to 30°C