LB-Agar, Powdered
LB-Agar, Powdered
Actual product may vary
MP Biomedicals
LB-Agar, Powdered
Offered in a variety of formats for convenience and ease of use
Have Questions?
Catalog NumberQuantityFormContent And Storage
MPB1130022311000 gPowder
Catalog number MPB113002231
Price (SGD)
623.00
EA
Add to cart
Quantity:
1000 g
Form:
Powder
Price (SGD)
623.00
EA
Add to cart
Specifications
Certifications/ComplianceMolecular biology Certified™ FDA
FormPowder
FormatCapsule
GradeMolecular biology
Product TypeAgar
FormulationTryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L, Agar 15g/L
For Use With (Application)For growth of bacteria
Quantity1000 g
Shelf Life4 Years
Unit SizeEA
Contents & Storage
15°C to 30°C