Product Image
MP Biomedicals
LB-Agar, Capsules, Molecular Biology Certified™
Rich medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium.
Have Questions?
Catalog NumberQuantityFormat
MPB113002221454 gCapsule
Catalog number MPB113002221
Price (SGD)
317.00
Each
Quantity:
454 g
Format:
Capsule
Price (SGD)
317.00
Each
  • Nutritionally rich medium originally developed for growth and maintenance of E. coli strains
  • Eliminates the waste, inaccuracies, and mess associated with weighing out bulk powder
  • Contains essential electrolytes for transport and osmotic balance, due to NaCl component
Specifications
Certifications/ComplianceMolecular Biology Certified™ FDA
FormatCapsule
GradeMolecular Biology
Product TypeAgar
FormulationTryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L, Agar 15g/L
For Use With (Application)For growth of bacteria
Quantity454 g
Shelf Life4 Years
Unit SizeEach
Contents & Storage
15°C to 30°C