▸
Microbiological Media and Media Additives
▸
Media Additives
▸
LB Medium, Large Capsules
Actual product may vary
MP Biomedicals
LB Medium, Large Capsules
Culture media used for the growth of bacteria.
Have Questions?
Change view
Catalog Number
Quantity
MPB113002046
5000 g
MPB113002036
1000 g
2 Options
Catalog number
MPB113002046
Price (SGD)
2,527.00
Each
Add to cart
Quantity:
5000 g
Price (SGD)
2,527.00
Each
Add to cart
Formulations for the
E. coli
growth media contain 10g of tryptone and 5g of yeast extract per liter, 10g NaCl per liter
Specifications
Certifications/Compliance
Molecular Biology Certified™
Format
Capsule
Product Type
Microbiology Media
Formulation
Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L
For Use With (Application)
For growth of bacteria
Quantity
5000 g
Unit Size
Each
Contents & Storage
15°C to 30°C