LB Medium, Dehydrated (Capsules)
LB Medium, Dehydrated (Capsules)
Actual product may vary
MP Biomedicals
LB Medium, Dehydrated (Capsules)
Formulations for the E. coli growth media known as LB Medium typically contain 10g of tryptone and 5g of yeast extract per liter, and LB Medium contains 10g NaCl per liter
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
MPB1130020415000 g
MPB113002021454 g
MPB1130020311000 g
Catalog number MPB113002041
Price (SGD)
3,425.00
EA
Add to cart
Quantity:
5000 g
Price (SGD)
3,425.00
EA
Add to cart
Specifications
Certifications/ComplianceMolecular Biology Certified™
FormDehydrated
Product TypeMicrobiology Media
FormulationTryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L
For Use With (Application)For growth of Escherichia coli, plasmid and other bacterial cultures in liquid medium
Quantity5000 g
Shelf Life4 Years
Unit SizeEA
Contents & Storage
15°C to 30°C