Check Order Status
L-Broth Agar, Capsules
Actual product may vary
MP Biomedicals
L-Broth Agar, Capsules
For the growth of
E.coli
and certified for plasmid and other bacterial cultures in liquid medium
Have Questions?
Catalog Number
Quantity
MPB113001221
454 g
Catalog number
MPB113001221
Price (SGD)
396.00
EA
Add to cart
Quantity:
454 g
Price (SGD)
396.00
EA
Add to cart
Specifications
Product Type
Agar
Sterility
Sterile
pH
6.5
Quantity
454 g
Unit Size
EA
Contents & Storage
15° to 30°C