Check Order Status
RNASE/DNASE-Free Water, MP Biomedicals™, 500mL
Element Search
Structure Search
MP Biomedicals
RNASE/DNASE-Free Water, MP Biomedicals™, 500mL
Have Questions?
Catalog Number
Quantity
MPB112450204
500 mL
Catalog number
MPB112450204
Price (SGD)
65.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
65.00
EA
Add to cart
Specifications
Chemical Name or Material
Water
Name Note
Not DEPC-treated.
CAS
7732-18-5
Color
Colorless
Molecular Formula
H2O
View more