ETBR Greenbag Disposal Kit
ETBR Greenbag Disposal Kit
Actual product may vary
MP Biomedicals
ETBR Greenbag Disposal Kit
ETBR Greenbag Disposal Kit for removing Ethidium Bromide from solutions.
Have Questions?
Catalog number MPB112350200
Price (SGD)
568.00
EA
Add to cart
Price (SGD)
568.00
EA
Add to cart
  • Each kit can remove 500 mg of Ethidium Bromide
  • Each bag contains a standard amount of activated material that will safely and easily concentrate Ethidium Bromide from large liquid quantities
  • The bags containing the biohazardous material can then be disposed of in a proper container
  • Applications include Agarose Gel Electrophoresis, DNA and RNA purification
  • Store at room temperature 15°C to 30°C
  • 50 bags/kit
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Specifications
DisposableSingle-use
For Use With (Application)Agarose gel electrophoresis, DNA, RNA purification
No. per Pack50 Bags/Pk.
Product TypeETBr Green Bag Disposal Kit
Unit SizeEA
Contents & Storage
Store at 15°C to 30°C