GeneClean II Kit
Actual product may vary
MP Biomedicals
GeneClean II Kit
GeneClean™ II Kit
Have Questions?
Catalog NumberNo. of Reactions
MPB111001400300 Preps
Catalog number MPB111001400
Price (SGD)
424.00
Each
No. of Reactions:
300 Preps
Price (SGD)
424.00
Each
  • The GENECLEAN™ II Kit is designed to optimally purify 200 bp to 20 kb DNA from TAE and TBE buffered agarose gels and solutions. Glassmilk™ slurry allows flexibility of reacion size and volume.
  • The purified DNA is suitable for a variety of downstream applications, such as sequencing, PCR, restriction digestion, cloning and labeling.
  • The GENECLEAN™ II Kit is designed for the rapid purification of linear and plasmid DNA fragments from standard or low-melting agarose gels.
  • This kit can also be used to purify DNA from polyacrylamide gels.
  • Typical recovery of DNA is up to 80%.
  • Each column can bind up to 10 μg of DNA from up to a 3.5 g agarose slice.
  • The GENECLEAN™ II Kit combines silica-binding technology with the convenience of a spin or vacuum column format.
  • DNA fragments of interest are extracted from slices of an agarose gel and are bound to a silica membrane.
  • Contaminants are removed by a simple spin or vacuum wash.
  • The bound DNA is then eluted.
  • DNA fragments of interest are extracted by solubilizing slices of an agarose gel.
  • The Gel Solubilization Solution dissolves an agarose gel slice from gels run in TBE or TAE buffer.
  • The extracted DNA fragments selectively adsorb onto the silica membrane in the presence of the Gel Solubilization Solution.
  • Contaminants are removed by a simple spin or vacuum wash. Finally, the bound DNA is eluted in Tris buffer.
Specifications
ConcentrationNew Wash/Ethanol Concentration must be ∽ 50%
For Use With (Application)Nucleic Acid Isolation & Purification
No. of Reactions300 Preps
pHAbove 500 bp: 4.5 M NaI pH 7.0 - 7.4; Between 200 - 500 bp: 4.5 M NaI pH 6.0; NEW Wash: pH Range: 7.4 - 8.0
Product TypeKits & Assays
SynonymRapid purification of linear DNA
FormSolution or Gels
Unit SizeEach
Contents & Storage
All gENECLEAN™ Kits are shipped at room temperature (15-30°C).